siRNA Details
Gene Symbol: Lce3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018079.1 | NP_001018089.1 | 2 | 571 | ||
GenBank mRNA | AK009081.1 | - | 1 | 423 | ||
BC060281.1 | - | 2 | 571 | 30288382 |

More Information
Other Species Targeted

Gene Ontology Categories:
