siRNA Details
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290662.1 | NP_001277591.1 | 13 | 1287 | ||
NM_134471.4 | NP_608301.3 | 13 | 1328 | |||
GenBank mRNA | AK150222.1 | - | 13 | 1259 | ||
AK161309.1 | - | 13 | 1287 | |||
BC006841.1 | - | 13 | 1288 | 3598157 |

More Information
Other Species Targeted

Gene Ontology Categories:
