siRNA Details
Gene Symbol: Arl4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177305.4 | NP_796279.2 | 1 | 680 | ||
GenBank mRNA | AK080335.1 | - | 1 | 624 | ||
AK080380.1 | - | 1 | 624 | |||
AK080743.1 | - | 1 | 633 | |||
BC049804.1 | - | 365 | ||||
BC055769.1 | - | 1 | 640 | 6414771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
