siRNA Details
Gene Symbol: Cetn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145825.2 | NP_665824.1 | 3 | 329 | ||
GenBank mRNA | AF362367.1 | - | 3 | 329 | ||
AK078275.1 | - | 2 | 2244 | |||
AK131808.1 | - | 3 | 317 | |||
AK166515.1 | - | 2 | 245 | |||
BC038154.1 | - | 2 | 220 | |||
BC060991.1 | - | 3 | 281 | 6742836 | ||
BC087905.1 | - | 3 | 307 | 6707456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
