siRNA Details
Gene Symbol: Mylk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297612.1 | NP_001284541.1 | 1 | 159 | ||
GenBank mRNA | AK165216.1 | - | 1 | 98 | ||
BB843262.1 | - | 1 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
