siRNA Details
Gene Symbol: Olfr287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011780.1 | NP_001011780.1 | 3 | 974 | ||
GenBank mRNA | AK140728.1 | - | 1 | 854 |
Gene Symbol: Olfr288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011733.2 | NP_001011733.2 | 3 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
