siRNA Details
Gene Symbol: Timm17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011590.2 | NP_035720.1 | 4 | 266 | ||
GenBank mRNA | AF106620.1 | - | 4 | 248 | ||
AK003114.1 | - | 4 | 273 | |||
AK003129.1 | - | 4 | 267 | |||
BC010830.1 | - | 4 | 254 | 4220707 | ||
BC098216.1 | - | 4 | 263 | 1527710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
