siRNA Details
Gene Symbol: Rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011029.4 | NP_035159.3 | 2, 3 | 240 | ||
GenBank mRNA | AF140348.1 | - | 2, 3 | 227 | ||
AK010423.1 | - | 2, 3 | 220 | |||
AK010985.1 | - | 2, 3 | 222 | |||
AK011041.1 | - | 2, 3 | 222 | |||
AK011043.1 | - | 2, 3 | 222 | |||
AK082935.1 | - | 2, 3 | 219 | |||
AK088954.1 | - | 2, 3 | 220 | |||
AK134224.1 | - | 2, 3 | 220 | |||
AK135488.1 | - | 2, 3 | 222 | |||
AK145713.1 | - | 2, 3 | 217 | |||
AK151123.1 | - | 2, 3 | 219 | |||
AK160551.1 | - | 2, 3 | 219 | |||
AK160625.1 | - | 2, 3 | 219 | |||
AK165219.1 | - | 2, 3 | 220 | |||
AK166697.1 | - | 2, 3 | 219 | |||
AK166874.1 | - | 2, 3 | 222 | |||
AK167132.1 | - | 2, 3 | 220 | |||
AK205896.1 | - | 1, 2 | 125 | |||
AK211265.1 | - | 1, 2 | 125 | |||
AK212093.1 | - | 1, 2 | 125 | |||
AK212679.1 | - | 1, 2 | 125 | |||
AK213565.1 | - | 1, 2 | 125 | |||
AK214711.1 | - | 1, 2 | 125 | |||
AK215661.1 | - | 1, 2 | 125 | |||
AK216125.1 | - | 1, 2 | 125 | |||
AK218136.1 | - | 1, 2 | 125 | |||
AK219743.1 | - | 1, 2 | 125 | |||
AK219820.1 | - | 1, 2 | 125 | |||
AV586849.1 | - | 2, 3 | 317 | |||
BC003829.1 | - | 2, 3 | 204 | 3600738 | ||
BC037195.1 | - | 1, 2 | 167 | 4500748 | ||
BC055886.1 | - | 1, 2 | 374 | 4240083 | ||
BC081461.1 | - | 1, 2 | 175 | 6820702 | ||
BC084677.1 | - | 2, 3 | 204 | 6476344 | ||
BC092041.1 | - | 2, 3 | 205 | 2651126 | ||
BC094902.1 | - | 2, 3 | 204 | 6433349 | ||
BC099601.1 | - | 2, 3 | 205 | 1245698 | ||
BC110285.1 | - | 2, 3 | 205 | 5151374 | ||
EU554633.1 | - | 1, 2 | 142 | |||
J02870.1 | - | 2, 3 | 221 | |||
X06406.1 | - | 2, 3 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
