siRNA Details
Gene Symbol: Chchd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025395.3 | NP_079671.1 | 2 | 212 | ||
GenBank mRNA | AK003341.1 | - | 2 | 114 | ||
AK003936.1 | - | 2 | 95 | |||
AK013018.1 | - | 2 | 91 | |||
BC094414.1 | - | 2 | 103 | 1515083 | ||
BC096668.1 | - | 2 | 110 | 1530928 | ||
BY335381.1 | - | 2 | 215 |

More Information
Other Species Targeted

Gene Ontology Categories:
