siRNA Details
Gene Symbol: 0610012G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002600.1 | - | 1 | 527 | ||
AK002881.1 | - | 1 | 492 | |||
AK009191.1 | - | 1 | 534 | |||
AK146135.1 | - | 1 | 548 | |||
BC021536.1 | - | 1 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
