siRNA Details
Gene Symbol: H2-DMb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169969.1 | - | 2 | 402 | ||
AK170866.1 | - | 2 | 404 | |||
BC002237.1 | - | 383 | 3491702 | |||
U20310.1 | - | 1 | 159 | |||
U20312.1 | - | 1 | 159 | |||
U35329.1 | - | 213 | ||||
U35330.1 | - | 213 | ||||
U35331.1 | - | 213 | ||||
U35332.1 | - | 213 | ||||
U35333.1 | - | 213 | ||||
X62743.1 | - | 321 | ||||
X90964.1 | - | 213 | ||||
X90965.1 | - | 213 | ||||
X90966.1 | - | 213 | ||||
X90967.1 | - | 213 | ||||
X90968.1 | - | 213 | ||||
X90969.1 | - | 213 | ||||
X90970.1 | - | 213 | ||||
X90971.1 | - | 213 | ||||
X90972.1 | - | 213 | ||||
X90973.1 | - | 213 | ||||
X90974.1 | - | 213 | ||||
X90975.1 | - | 213 |
