siRNA Details
Gene Symbol: Rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013725.4 | NP_038753.1 | 2 | 161 | ||
GenBank mRNA | AK005147.1 | - | 2 | 106 | ||
AK150738.1 | - | 2 | 136 | |||
BC012641.1 | - | 2 | 175 | 4019309 | ||
CJ044201.1 | - | 2 | 162 | |||
U93864.1 | - | 2 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
