siRNA Details
Gene Symbol: Skor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109743.1 | NP_001103213.1 | 6 | 2964 | ||
GenBank mRNA | AB358976.1 | - | 6 | 2964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
