siRNA Details
Gene Symbol: Slfn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172796.2 | NP_766384.2 | 2 | 1244 | ||
GenBank mRNA | AK036579.1 | - | 2 | 1245 | ||
AK050355.1 | - | 2 | 1198 | |||
AY217035.1 | - | 1 | 712 | |||
AY261800.1 | - | 2 | 1152 | |||
AY261801.1 | - | 2 | 1134 |
Gene Symbol: Slfn10-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041394.1 | - | 1 | 485 | ||
AK144698.1 | - | 2 | 1078 | |||
AK220241.1 | - | 2 | 1202 | |||
AY261802.1 | - | 2 | 1077 | |||
AY261803.1 | - | 2 | 1046 |
Gene Symbol: Slfn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167743.1 | NP_001161215.1 | 2 | 1147 | ||
NM_181545.4 | NP_853523.2 | 3 | 1351 | |||
GenBank mRNA | AK150875.1 | - | 1 | 673 | ||
AY261798.1 | - | 3 | 1351 | |||
BC152549.1 | - | 2 | 1147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
