siRNA Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022565.2 | NP_072087.2 | 2 | 1612 | ||
GenBank mRNA | AB036838.1 | - | 1 | 1062 | ||
AK140992.1 | - | 2 | 1613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
