siRNA Details
Gene Symbol: Mrps22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025485.3 | NP_079761.1 | 8 | 1051 | ||
GenBank mRNA | AK003836.1 | - | 7 | 819 | ||
AK013925.1 | - | 8 | 1032 | |||
AK154600.1 | - | 8 | 1032 | |||
BC021882.1 | - | 3 | 270 | |||
BC051198.1 | - | 8 | 1031 | 1396237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
