siRNA Details
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026080.2 | NP_080356.1 | 4 | 943 | ||
GenBank mRNA | AK014099.1 | - | 4 | 930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
