siRNA Details
Gene Symbol: Xpo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023045.2 | NP_075532.1 | 6 | 677 | ||
GenBank mRNA | AJ297360.1 | - | 6 | 583 | ||
AK028303.1 | - | 4 | 422 | |||
AK035775.1 | - | 6 | 602 | |||
AK036837.1 | - | 4 | 422 | |||
AK051105.1 | - | 6 | 599 | |||
AK122361.1 | - | 5 | 562 | |||
AK143729.1 | - | 6 | 646 | |||
AK164595.1 | - | 6 | 605 | |||
BC029702.1 | - | 6 | 603 | 5053250 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108386, XM_008770832, XM_006252282.
Gene Ontology Categories:
