siRNA Details
Gene Symbol: Dear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040461.2 | NP_001035551.2 | 1 | 475 | ||
GenBank mRNA | BC141214.1 | - | 1 | 467 | 9055826 | |
DQ009865.1 | - | 1 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
