siRNA Details
Gene Symbol: Defb25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039122.1 | NP_001034211.1 | 2 | 254 | ||
GenBank mRNA | BC132662.1 | - | 2 | 250 | 40130939 | |
DQ012036.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
