siRNA Details
Gene Symbol: Gm15386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040027.2 | NP_001035116.1 | 2 | 129 | ||
GenBank mRNA | DQ012028.1 | - | 2 | 71 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
