siRNA Details
Gene Symbol: Defb47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039125.2 | NP_001034214.1 | 2 | 199 | ||
GenBank mRNA | BC132604.1 | - | 2 | 199 | 40130881 | |
BC132606.1 | - | 2 | 199 | 40130883 | ||
DQ012043.1 | - | 2 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
