siRNA Details
Gene Symbol: Tmem176a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098271.1 | NP_001091741.1 | 3 | 300 | ||
NM_025326.4 | NP_079602.4 | 3 | 352 | |||
GenBank mRNA | AB063313.1 | - | 2 | 242 | ||
AK002546.1 | - | 3 | 328 | |||
AK004315.1 | - | 3 | 325 | |||
AK079378.1 | - | 314 | ||||
AK150576.1 | - | 3 | 317 | |||
AK162991.1 | - | 2 | 474 | |||
AK165660.1 | - | 3 | 326 | |||
BC010831.1 | - | 3 | 296 | 4221626 | ||
BY018646.1 | - | 3 | 369 | |||
BY352519.1 | - | 3 | 303 | |||
CT010190.1 | - | 2 | 242 |
