siRNA Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025364.2 | NP_079640.1 | 5 | 453 | ||
GenBank mRNA | AK003453.1 | - | 5 | 305 | ||
AK018773.1 | - | 5 | 316 | |||
AK088266.1 | - | 5 | 302 | |||
AK146248.1 | - | 5 | 331 | |||
AK212934.1 | - | 6 | 337 | |||
BC027510.1 | - | 5 | 292 | 1382354 | ||
BC094393.1 | - | 5 | 289 | |||
BC157909.1 | - | 5 | 317 | 9088111 | ||
BC158023.1 | - | 5 | 317 | 9007329 | ||
BC172063.1 | - | 5 | 314 | 9054349 |
