siRNA Details
Gene Symbol: Cxx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018063.1 | NP_001018073.1 | 1 | 508 | ||
GenBank mRNA | AK003646.1 | - | 1 | 487 | ||
BC029838.1 | - | 1 | 508 | 3967024 |
Gene Symbol: Cxx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024170.2 | NP_077132.1 | 1 | 540 | ||
GenBank mRNA | AK002323.1 | - | 1 | 476 | ||
BC003436.1 | - | 1 | 459 | 2647372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
