siRNA Details
Gene Symbol: Fam53c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175104.4 | NP_780313.1 | 5 | 2193 | ||
GenBank mRNA | AK044469.1 | - | 5 | 1997 | ||
AK135712.1 | - | 4 | 1939 | |||
AK154214.1 | - | 5 | 1997 | |||
AK166841.1 | - | 4 | 1937 | |||
BC044896.1 | - | 4 | 1918 | |||
BC057111.1 | - | 5 | 2193 | 6417613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
