siRNA Details
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039959.2 | NP_001035048.1 | 4 | 480 | ||
NM_001286518.1 | NP_001273447.1 | 4 | 480 | |||
NM_009643.2 | NP_033773.1 | 4 | 480 | |||
GenBank mRNA | AK009866.1 | - | 4 | 503 | ||
AK045184.1 | - | 4 | 481 | |||
AK134304.1 | - | 4 | 1492 | |||
BC028439.1 | - | 3 | 345 | |||
CB321491.1 | - | 4 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
