siRNA Details
Gene Symbol: Ndufa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023202.4 | NP_075691.1 | 4 | 390 | ||
GenBank mRNA | AK002476.1 | - | 4 | 371 | ||
AK003468.1 | - | 4 | 371 | |||
AK010299.1 | - | 4 | 348 | |||
BC052817.1 | - | 4 | 354 | 6487401 | ||
BC055698.1 | - | 4 | 355 | 6411262 | ||
BX518947.1 | - | 3 | 267 | |||
BX522503.1 | - | 2 | 118 | |||
KJ650204.1 | - | 4 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
