siRNA Details
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 2 | 459 | ||
NM_023210.4 | NP_075699.3 | 2 | 459 | |||
GenBank mRNA | AB037685.1 | - | 2 | 134 | ||
AK012759.1 | - | 2 | 426 | |||
AK049647.1 | - | 2 | 426 | |||
AK076049.1 | - | 2 | 286 | |||
AK088401.1 | - | 2 | 439 | |||
AK147888.1 | - | 2 | 425 | |||
AK168380.1 | - | 2 | 408 | |||
AK172146.1 | - | 2 | 473 | |||
AK205819.1 | - | 1 | 73 | |||
BC005690.1 | - | 2 | 394 | 3596597 | ||
BC080684.1 | - | 2 | 399 | 6493476 | ||
CF952181.1 | - | 2 | 130 | |||
U89345.2 | - | 2 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
