siRNA Details
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023211.4 | NP_075700.2 | 3 | 219 | ||
GenBank mRNA | AK008345.1 | - | 3 | 220 | ||
AK008511.1 | - | 1049 | ||||
AK038457.1 | - | 3 | 226 | |||
AK088856.1 | - | 3 | 237 | |||
BC024355.1 | - | 3 | 300 | |||
BC087919.1 | - | 3 | 219 | 6545418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
