siRNA Details
Gene Symbol: A430089I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177913.4 | NP_808581.1 | 4 | 1241 | ||
GenBank mRNA | AK040373.1 | - | 4 | 1242 | ||
BC147369.1 | - | 4 | 1208 | 9056433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
