siRNA Details
Gene Symbol: Ndufb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025597.2 | NP_079873.1 | 3 | 430 | ||
GenBank mRNA | AK007930.1 | - | 3 | 429 | ||
AK009008.1 | - | 3 | 392 | |||
AK012320.1 | - | 3 | 431 | |||
AK013608.1 | - | 3 | 416 | |||
AK138123.1 | - | 3 | 428 | |||
BC028669.1 | - | 3 | 586 | |||
BC036989.1 | - | 3 | 427 | 4989233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
