siRNA Details
Gene Symbol: Dda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294259.1 | NP_001281188.1 | 3, 4 | 244 | ||
NM_025600.2 | NP_079876.1 | 4, 5 | 325 | |||
GenBank mRNA | AK006318.1 | - | 4, 5 | 309 | ||
AK007085.1 | - | 4, 5 | 344 | |||
AK032966.1 | - | 4, 5 | 283 | |||
AK149117.1 | - | 3, 4 | 255 | |||
AK149980.1 | - | 4, 5 | 305 | |||
AK160132.1 | - | 4, 5 | 322 | |||
AK168085.1 | - | 4, 5 | 283 | |||
BB610194.1 | - | 4, 5 | 318 | |||
BC034701.1 | - | 4, 5 | 285 | 3494430 | ||
BC093523.1 | - | 4, 5 | 268 | 3711509 | ||
BY297077.1 | - | 3, 4 | 204 | |||
BY316490.1 | - | 4, 5 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
