siRNA Details
Gene Symbol: Gm7457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039138.1 | - | 1, 2 | 354 | ||
AK143902.1 | - | 2, 3 | 382 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039138.1 | - | 1, 2 | 354 | ||
AK143902.1 | - | 2, 3 | 382 |