siRNA Details
Gene Symbol: Chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293776.1 | NP_001280705.1 | 4 | 646 | ||
NM_001293777.1 | NP_001280706.1 | 4 | 643 | |||
NM_001293778.1 | NP_001280707.1 | 4 | 646 | |||
NM_001293779.1 | NP_001280708.1 | 4 | 643 | |||
NM_001293780.1 | NP_001280709.1 | 3 | 1929 | |||
NM_001293781.1 | NP_001280710.1 | 3 | 1926 | |||
NM_001293782.1 | NP_001280711.1 | 4 | 1643 | |||
NM_023215.6 | NP_075704.2 | 3 | 1929 | |||
GenBank mRNA | AB041654.1 | - | 3 | 1881 | ||
AK004558.2 | - | 3 | 1875 | |||
AK005377.1 | - | 4 | 608 | |||
AK010866.1 | - | 4 | 628 | |||
AK047914.1 | - | 4 | 620 | |||
AK077960.1 | - | 3 | 1915 | |||
AK132769.1 | - | 4 | 619 | |||
AK149010.1 | - | 1 | 117 | |||
AK152492.1 | - | 4 | 599 | |||
AK190161.1 | - | 4 | 358 | |||
AK204756.1 | - | 3 | 354 | |||
BC051939.1 | - | 4 | 1597 | |||
BC066837.1 | - | 3 | 1882 | |||
BC083333.1 | - | 2 | 232 | |||
BC108396.1 | - | 3 | 275 | |||
BC120651.1 | - | 2 | 1166 | 40129574 | ||
BC144775.1 | - | 2 | 1166 | 9053323 | ||
BQ746440.1 | - | 4 | 600 | |||
CX234428.1 | - | 4 | 486 |

More Information
Other Species Targeted

Gene Ontology Categories:
