siRNA Details
Gene Symbol: Adamtsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008355.1 | - | 8 | 918 | ||
AK008364.1 | - | 8 | 918 | |||
AK008440.1 | - | 8 | 917 | |||
AK029081.1 | - | 8 | 894 | |||
AK154947.1 | - | 5 | 543 | |||
BC058577.1 | - | 4 | 1529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
