siRNA Details
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 5 | 586 | ||
GenBank mRNA | AK002714.1 | - | 4 | 532 | ||
AK010619.1 | - | 5 | 529 | |||
AK150956.1 | - | 5 | 531 | |||
AK151073.1 | - | 5 | 531 | |||
AK152933.1 | - | 5 | 533 | |||
AK160490.1 | - | 5 | 525 | |||
AK168248.1 | - | 5 | 534 | |||
AK168656.1 | - | 5 | 525 | |||
AK190324.1 | - | 2 | 213 | |||
BC083354.1 | - | 4 | 508 | 5715690 | ||
BC089034.1 | - | 4 | 510 | 30675988 | ||
X78682.1 | - | 4 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
