siRNA Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025633.3 | NP_079909.1 | 2 | 120 | ||
GenBank mRNA | AK005241.1 | - | 2 | 117 | ||
AK010067.1 | - | 2 | 120 | |||
BC060739.1 | - | 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
