siRNA Details
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006166.1 | - | 1 | 90 | ||
AK011393.1 | - | 1 | 84 | |||
AK011848.1 | - | 1 | 90 | |||
AK017100.1 | - | 1 | 121 | |||
AK017322.1 | - | 1 | 84 | |||
AK029148.1 | - | 86 | ||||
AK030275.1 | - | 84 | ||||
AK147270.1 | - | 97 | ||||
AK170765.1 | - | 1 | 90 | |||
BC026913.1 | - | 1 | 85 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
