siRNA Details
Gene Symbol: Ribc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025660.2 | NP_079936.1 | 6 | 841 | ||
GenBank mRNA | AK011592.1 | - | 6 | 841 | ||
AK017461.1 | - | 6 | 733 | |||
BC050755.1 | - | 6 | 829 | 6772058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
