siRNA Details
Gene Symbol: Zscan25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081431.1 | NP_001074900.1 | 1 | 178 | ||
GenBank mRNA | BC150878.1 | - | 1 | 169 | 9087789 | |
BC151110.1 | - | 1 | 169 | 9088023 | ||
BC172035.1 | - | 1 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
