siRNA Details
Gene Symbol: Snrpg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026506.2 | NP_080782.1 | 3 | 164 | ||
GenBank mRNA | AK011129.1 | - | 3 | 163 | ||
AK012802.1 | - | 3 | 153 | |||
AK013235.1 | - | 3 | 165 | |||
AK017852.1 | - | 3 | 513 | |||
BC027499.1 | - | 3 | 137 | 1430048 | ||
BC051470.1 | - | 3 | 209 | 1244608 | ||
BC094411.1 | - | 3 | 151 | 1380524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
