siRNA Details
Gene Symbol: Tceal8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168578.1 | NP_001162050.1 | 4 | 559 | ||
NM_025703.3 | NP_079979.2 | 3 | 457 | |||
GenBank mRNA | AK012030.1 | - | 3 | 382 | ||
AK014459.1 | - | 3 | 392 | |||
AK027927.1 | - | 3 | 385 | |||
AK076203.1 | - | 3 | 432 | |||
AK083347.1 | - | 3 | 385 | |||
AK137039.1 | - | 3 | 379 | |||
AK156414.1 | - | 3 | 385 | |||
BC027269.1 | - | 3 | 411 | 3588435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
