siRNA Details
Gene Symbol: Gm8369 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164202.1 | NP_001157674.1 | 4, 5 | 384 | ||
GenBank mRNA | BB637086.1 | - | 5, 6 | 517 | ||
BE861164.1 | - | 3, 4 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
