siRNA Details
Gene Symbol: Vmn1r151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166712.1 | NP_001160184.1 | 1 | 878 |
Gene Symbol: Vmn1r103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166737.1 | NP_001160209.1 | 1 | 878 |
