siRNA Details
Gene Symbol: Gm8765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244649.1 | NP_001231578.1 | 4 | 725 | ||
GenBank mRNA | BC172081.1 | - | 4 | 725 | 9054367 |
Gene Symbol: Gm906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033438.2 | NP_001028610.1 | 4 | 742 | ||
GenBank mRNA | AK133043.1 | - | 4 | 743 | ||
BC147515.1 | - | 4 | 725 | 9056507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
