siRNA Details
Gene Symbol: Pgam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023418.2 | NP_075907.2 | 4 | 820 | ||
GenBank mRNA | AF283667.1 | - | 4 | 732 | ||
AK004921.1 | - | 4 | 676 | |||
AK009905.2 | - | 4 | 673 | |||
AK144681.1 | - | 4 | 674 | |||
AK150716.1 | - | 4 | 669 | |||
AK152443.1 | - | 4 | 669 | |||
AK153202.1 | - | 4 | 673 | |||
AK159305.1 | - | 4 | 673 | |||
AK165410.1 | - | 4 | 669 | |||
AK168949.1 | - | 4 | 666 | |||
AK215282.1 | - | 2 | 92 | |||
AK218196.1 | - | 2 | 92 | |||
AK219760.1 | - | 2 | 92 | |||
BC002241.1 | - | 4 | 768 | 3491964 | ||
BC005661.1 | - | 4 | 734 | 3588761 | ||
BC066844.1 | - | 4 | 657 | 30471755 | ||
BC067412.1 | - | 3 | 473 | |||
BC083090.1 | - | 4 | 669 | 30444229 | ||
BC106139.1 | - | 4 | 740 | 6528521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
