siRNA Details
Gene Symbol: Rps3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016959.4 | NP_058655.3 | 2 | 173 | ||
GenBank mRNA | AK003158.1 | - | 2 | 99 | ||
AK010605.1 | - | 2 | 96 | |||
AK012345.1 | - | 2 | 96 | |||
AK050610.1 | - | 2 | 99 | |||
AK088113.1 | - | 2 | 96 | |||
AK135318.1 | - | 2 | 100 | |||
AK146304.1 | - | 2 | 107 | |||
AK148513.1 | - | 2 | 99 | |||
AK150441.1 | - | 2 | 99 | |||
AK150796.1 | - | 2 | 109 | |||
AK150814.1 | - | 2 | 99 | |||
AK151429.1 | - | 2 | 100 | |||
AK151982.1 | - | 2 | 109 | |||
AK159777.1 | - | 2 | 99 | |||
AK166713.1 | - | 2 | 97 | |||
AK167512.1 | - | 2 | 100 | |||
AK167562.1 | - | 2 | 98 | |||
AK168240.1 | - | 2 | 97 | |||
AK190515.1 | - | 2 | 70 | |||
AK196258.1 | - | 2 | 85 | |||
AK200497.1 | - | 2 | 70 | |||
AK207259.1 | - | 2 | 70 | |||
AK207595.1 | - | 2 | 62 | |||
AK207749.1 | - | 2 | 46 | |||
AK208520.1 | - | 2 | 70 | |||
AK208628.1 | - | 2 | 70 | |||
AL118161.1 | - | 2 | 178 | |||
BC039659.1 | - | 2 | 98 | 5025347 | ||
BC081451.1 | - | 2 | 86 | 6824188 | ||
BC083338.1 | - | 2 | 79 | 6820218 | ||
BC084675.1 | - | 2 | 90 | 5319512 | ||
BC106115.1 | - | 2 | 88 | 2599128 | ||
M88335.1 | - | 2 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
