siRNA Details
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026533.3 | NP_080809.1 | 4, 5 | 408 | ||
GenBank mRNA | AK012479.1 | - | 4, 5 | 353 | ||
AK018703.1 | - | 4, 5 | 352 | |||
AK076060.1 | - | 4, 5 | 352 | |||
AK088237.1 | - | 3, 4 | 985 | |||
AK151043.1 | - | 4, 5 | 353 | |||
AK151671.1 | - | 4, 5 | 354 | |||
AK166691.1 | - | 4, 5 | 353 | |||
AK177445.1 | - | 351 | ||||
AV498110.1 | - | 3, 4 | 315 | |||
BC011192.1 | - | 4, 5 | 351 | 4016679 | ||
BC090397.1 | - | 4, 5 | 335 | 5150054 | ||
BC106137.1 | - | 4, 5 | 336 | 4990874 | ||
BC125602.1 | - | 4, 5 | 352 | 40130117 | ||
BC125604.1 | - | 4, 5 | 352 | 40130119 |

More Information
Other Species Targeted

Gene Ontology Categories:
