siRNA Details
Gene Symbol: Scgb2b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281508.1 | NP_001268437.1 | 2 | 154 | ||
GenBank mRNA | CK616067.1 | - | 2 | 162 |
Gene Symbol: Scgb2b19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199336.1 | NP_001186265.1 | 2 | 160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
